Läs mer
D-Lyxono-1,4-lactone, TRC
Produktkod. 30468743
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30468743 10 g
1 options
Produktkod. 30468743 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL49000010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material D-Lyxono-1,4-lactone
CAS 15384-34-6
Molekylformel C5 H8 O5
Kvantitet 10 g
Synonym D-Lyxonic acid, γ-lactone, Lyxonic acid, γ-lactone, D- (8CI), D-Lyxono-1,4-lactone, D-Lyxono-γ-lactone, Lyxonic acid-1,4-lactone, D-Lyxonolactone
InChI formel InChI=1S/C5H8O5/c6-1-2-3(7)4(8)5(9)10-2/h2-4,6-8H,1H2/t2-,3+,4+/m1/s1
LEDER OC[C@H]1OC(=O)[C@@H](O)[C@H]1O
IUPAC-namn (3S,4R,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-one
Molekylvikt (g/mol) 148.11
Formel vikt 148.0372