missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(+)-Maltose Monohydrate, ˜99%, MP Biomedicals™
Click to view available options
Quantity:
100 g
500 g
1 kg
Description
- D-(+)-Maltose monohydrate is used as a sweetener with about one-third the sweetness of sucrose and as a nutrient in culture media.
- It is used in pharmaceutical formulations and as a parenteral supplement of sugar for diabetics.
Specifications
Color | White |
Density | 1.54 g/mL |
Assay Percent Range | 99% |
Quantity | 100 g |
Optical Rotation | 130° ± 5°(c=4, H2O, NH3OH) |
Formula Weight | 360.31 |
Percent Purity | 99% |
Physical Form | Powder |
Chemical Name or Material | D-(+)-Maltose monohydrate, Molecular biology |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'