Läs mer
D-Mannoheptulose, TRC
Produktkod. 30012864
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012864 5 mg
30006408 25 mg
30004192 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012864 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM1651005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3615-44-9
Molekylformel C7 H14 O7
Molekylvikt (g/mol) 210.18
Synonym D-Manno-2-heptulose, (+)-Mannoheptulose, NSC 226836;
IUPAC-namn (3S,4S,5S,6R)-2,6-bis(hydroxymethyl)oxane-2,3,4,5-tetrol
LEDER OC[C@H]1OC(O)(CO)[C@@H](O)[C@@H](O)[C@@H]1O
Specifikationer
Kemiskt namn eller material D-Mannoheptulose
Kvantitet 5 mg
InChI formel InChI=1S/C7H14O7/c8-1-3-4(10)5(11)6(12)7(13,2-9)14-3/h3-6,8-13H,1-2H2/t3-,4-,5+,6+,7?/m1/s1
Formel vikt 210.074