Läs mer
D-Mannose-6-O-sulphate Sodium Salt, TRC
Produktkod. 30179086
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179086 10 mg
30179196 5 mg
30179492 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179086 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM18501310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 204575-08-6
Molekylformel C6H11O9SNa
Molekylvikt (g/mol) 282.2
Synonym D-Mannose, 6-(hydrogen sulfate), monosodium salt (9CI), D-Mannose 6-(hydrogen sulfate) sodium salt (1:1), D-Mannose-6-O-sulfate sodium salt, Sodium D-Mannose 6-O-sulfate
IUPAC-namn sodium;[(2R,3S,4S,5S)-3,4,5,6-tetrahydroxyoxan-2-yl]methyl sulfate
LEDER [Na+].OC1O[C@H](COS(=O)(=O)[O-])[C@@H](O)[C@H](O)[C@@H]1O
Specifikationer
Kemiskt namn eller material D-Mannose-6-O-sulphate Sodium Salt
Kvantitet 10 mg
InChI formel InChI=1S/C6H12O9S.Na/c7-3-2(1-14-16(11,12)13)15-6(10)5(9)4(3)8;/h2-10H,1H2,(H,11,12,13);/q;+1/p-1/t2-,3-,4+,5+,6?;/m1./s1
Formel vikt 282.0021