Läs mer
D-Melphalan, TRC
Produktkod. 30468122
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30468122 10 mg
1 options
Produktkod. 30468122 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM21691510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material D-Melphalan
CAS 13045-94-8
Molekylformel C13H18Cl2N2O2
Kvantitet 10 mg
Synonym 4-[Bis(2-chloroethyl)amino]-D-phenylalanine, CB 3026, D-Melphalan, D-Phenylalanine Mustard, Medphalan, NSC 35051
InChI formel InChI=1S/C13H18Cl2N2O2/c14-5-7-17(8-6-15)11-3-1-10(2-4-11)9-12(16)13(18)19/h1-4,12H,5-9,16H2,(H,18,19)/t12-/m1/s1
LEDER N[C@H](CC1=CC=C(N(CCCl)CCCl)C=C1)C(O)=O
Molekylvikt (g/mol) 305.2