missing translation for 'onlineSavingsMsg'
Learn More

D-myo-Inositol-1,4,5-trisphosphate sodium salt, 98%, Thermo Scientific Chemicals

Catalog Number 15498609
Click to view available options
Quantity:
0.1 mg
0.5 mg
1 mg
This item is not returnable. View return policy
MFCD00055051
Ins(1,4,5)P3 sodium salt; 1,4,5-IP3 sodium salt
This item is not returnable. View return policy
MFCD00055051
Ins(1,4,5)P3 sodium salt; 1,4,5-IP3 sodium salt

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 0.1 mg
Formula Weight 486.04
Percent Purity 98%
Physical Form Lyophilized Powder
Chemical Name or Material D-myo-Inositol-1,4,5-trisphosphate sodium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'