Learn More
D(-)-N-Methylglucamine, Sigma-Aldrich™
Product Code. 15834401
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
Product Code. Quantity Unit Size
15834401 250 g 250g
1 options
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15834401 Supplier Sigma Aldrich Supplier No. 8183180250
 Restricted Item
Beräknad leverans: 19-11-2026
MQ200 Graded - For Research and Non-regulated Industrial Applications
For resolution of racemates for synthesis
TRUSTED_SUSTAINABILITY
Specifications
CAS 6284-40-8
Melting Point 129°C to 131.5°C (Lit.)
Boiling Point 210°C/1013 hPa (Decomposes)
MDL Number MFCD00004707
Quantity 250 g
Synonym N-Methyl-ᴅ-glucamine, 1-Deoxy-1-methylaminosorbitol, 1-Deoxy-1-(methylamino)-ᴅ-glucitol, Meglumine, N-Methyl-ᴅ-glucamine, 1-Deoxy-1-(methylamino)-ᴅ-glucitol, Meglumine
Molecular Weight (g/mol) 195.21
Grade Research