Läs mer
D-(-)-Norgestrel, TRC
Produktkod. 30011952
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011952 100 mg
30007621 50 mg
30011850 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011952 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN689510100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 797-63-7
Molekylvikt (g/mol) 312.45
Synonym Levonorgestrel, (17α)-13-Ethyl-17-hydroxy-18,19-dinorpregn-4-en-20-yn-3-one, 13-Ethyl-17-hydroxy-18,19-dinor-17α-pregn-4-en-20-yn-3-one, (-)-Norgestrel, 13-Ethyl-17-ethynyl-17β-hydroxy-4-gonen-3-one, 13-Ethyl-17-hydroxy-18,19-dinor-17α-pregn-4-en-20-yn-3-one, 13-Ethyl-17α-ethynyl-17-hydroxygon-4-en-3-one, 13-Ethyl-17α-ethynylgon-4-en-17β-ol-3-one, 13β-Ethyl-17α-ethynyl-17β-hydroxygon-4-en-3-one, 17α-Ethynyl-13-ethyl-19-nortestosterone, 17α-Ethynyl-13β-ethyl-3-oxo-4-estren-17β-ol, 17α-Ethynyl-17-hydroxy-18-methylestr-4-en-3-one, 17α-Ethynyl-18-homo-19-nortestosterone, 17α-Ethynyl-18-methyl-19-nortestosterone, 17β-Hydroxy-18-methyl-19-nor-17α-pregn-4-en-20-yn-3-one, 18-Methyl-17α-ethynyl-19-nortestosterone, 18-Methylnorethindrone, 19-Nortestosterone, 17-ethynyl-18-methyl-, D-(-)-Norgestrel, D-Norgestrel, Dexnorgestrel, Femilis, Femilis Slim, Follistrel, Jadelle, Levonorgestrel, Levonova, Lng-IUS, Microlut, Microluton, Microval, Mirena, Norgeston, Norgestrelem, Norlevo, Norplant, Norplant 2, Norplant II, Postinor 2, Wy 5104, d-(17α)-17-Hydroxy-13-ethyl-18,19-dinorpregn-4-en-20-yn-3-one
LEDER CC[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1CC[C@@]2(O)C#C
Specifikationer
Kemiskt namn eller material D-(-)-Norgestrel
Anteckningar om renhetsgrad HPLC
Molekylformel C21H28O2
Kvantitet 100 mg
InChI formel InChI=1S/C21H28O2/c1-3-20-11-9-17-16-8-6-15(22)13-14(16)5-7-18(17)19(20)10-12-21(20,23)4-2/h2,13,16-19,23H,3,5-12H2,1H3/t16-,17+,18+,19-,20-,21-/m0/s1
Formel vikt 312.2089
Procent renhet >95%