Läs mer
D-o-Tyrosine, TRC
Produktkod. 30450501
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30450501 50 mg
1 options
Produktkod. 30450501 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT89998550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material D-o-Tyrosine
CAS 24008-77-3
Molekylformel C9 H11 N O3
Kvantitet 50 mg
Synonym D-Phenylalanine','2-hydroxy-, o-Tyrosine','D- (8CI), 2-Hydroxy-D-phenylalanine, (+)-o-Tyrosine, (2R)-2-Amino-3-(2-hydroxyphenyl)propanoic acid, (R)-2-Amino-3-(2-hydroxyphenyl)propanoic acid, (R)-2-Amino-3-(2-hydroxyphenyl)propionic acid, (r)-2-Amino-3-(2-hydroxyphenyl)propanoic acid, D-o-Hydroxyphenylalanine, D-o-Tyrosine, D-α-Amino-β-(o-hydroxyphenyl)propionic acid
InChI formel InChI=1S/C9H11NO3/c10-7(9(12)13)5-6-3-1-2-4-8(6)11/h1-4,7,11H,5,10H2,(H,12,13)/t7-/m1/s1
LEDER N[C@H](Cc1ccccc1O)C(=O)O
IUPAC-namn (2R)-2-amino-3-(2-hydroxyphenyl)propanoic acid
Molekylvikt (g/mol) 181.19
Formel vikt 181.0739