Läs mer
D-(+)-Octopine, TRC
Produktkod. 30012732
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012732 500 mg
30004457 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012732 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO239850500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 34522-32-2
Molekylformel C9 H18 N4 O4
Molekylvikt (g/mol) 246.26
Synonym N2-[(1R)-1-Carboxyethyl]-L-arginine, (R)-N2-(1-carboxyethyl)-L-arginine, Octopine, D-Octopine;
IUPAC-namn (2S)-5-carbamimidamido-2-[[(1R)-1-carboxyethyl]amino]pentanoic acid
LEDER C[C@@H](N[C@@H](CCCNC(=N)N)C(=O)O)C(=O)O
Specifikationer
Kemiskt namn eller material D-(+)-Octopine
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C9H18N4O4/c1-5(7(14)15)13-6(8(16)17)3-2-4-12-9(10)11/h5-6,13H,2-4H2,1H3,(H,14,15)(H,16,17)(H4,10,11,12)/t5-,6+/m1/s1
Kvantitet 500 mg
Formel vikt 246.1328
Procent renhet >95%