Läs mer
D-p-Hydroxyphenylglycinamide (>85%), TRC
Produktkod. 30406872
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30406872 25 mg
1 options
Produktkod. 30406872 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH95132025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2R)-2-Amino-2-(4-hydroxyphenyl)acetamide
CAS 54397-23-8
Molekylformel C8 H10 N2 O2
Kvantitet 25 mg
Synonym (2R)-2-Amino-2-(4-hydroxyphenyl)acetamide, Benzeneacetamide, α-amino-4-hydroxy-, (αR)-, Benzeneacetamide, α-amino-4-hydroxy-, (R)-, (αR)-α-Amino-4-hydroxybenzeneacetamide, (R)-2-Amino-2-(4-hydroxyphenyl)acetamide, (R)-α-Amino-4-hydroxybenzeneacetamide, 4-Hydroxy-(R)-phenylglycinamide, D-p-Hydroxyphenylglycinamide, D-α-Amino-p-hydroxyphenylacetamide
InChI formel InChI=1S/C8H10N2O2/c9-7(8(10)12)5-1-3-6(11)4-2-5/h1-4,7,11H,9H2,(H2,10,12)/t7-/m1/s1
LEDER N[C@@H](C(=O)N)c1ccc(O)cc1
IUPAC-namn (2R)-2-amino-2-(4-hydroxyphenyl)acetamide
Molekylvikt (g/mol) 166.18
Formel vikt 166.0742