Läs mer
D-Pantethine, TRC
Produktkod. 30409239
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30409239 100 mg
1 options
Produktkod. 30409239 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP205823100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material D-Pantethine
CAS 16816-67-4
Molekylformel C22 H42 N4 O8 S2
Kvantitet 100 mg
Synonym Butanamide','N,N'-[dithiobis[2,1-ethanediylimino(3-oxo-3,1-propanediyl)]]bis[2,4-dihydroxy-3,3-dimethyl-','(2R,2'R)-, Butanamide','N,N'-[dithiobis[2,1-ethanediylimino(3-oxo-3,1-propanediyl)]]bis[2,4-dihydroxy-3,3-dimethyl-','[R-(R*,R*)]-, Butyramide','N,N'-[dithiobis(ethyleneiminocarbonylethylene)]bis[2,4-dihydroxy-3,3-dimethyl-','D-(+)- (8CI), (2R,2'R)-N,N'-[Dithiobis[2,1-ethanediylimino(3-oxo-3,1-propanediyl)]]bis[2,4-dihydroxy-3,3-dimethylbutanamide], Bis(N-pantothenylamidoethyl) disulfide, Bis(pantothenamidoethyl) disulfide, D-Bis-pantothenyl-β-aminoethyl disulfide, D-Pantethine, LBF disulfide, Lipodel, Pantethine, Pantetina, Panthecin, Pantomin, Pantosin
InChI formel InChI=1S/C22H42N4O8S2/c1-21(2,13-27)17(31)19(33)25-7-5-15(29)23-9-11-35-36-12-10-24-16(30)6-8-26-20(34)18(32)22(3,4)14-28/h17-18,27-28,31-32H,5-14H2,1-4H3,(H,23,29)(H,24,30)(H,25,33)(H,26,34)/t17-,18-/m0/s1
LEDER CC(C)(CO)[C@@H](O)C(=O)NCCC(=O)NCCSSCCNC(=O)CCNC(=O)[C@H](O)C(C)(C)CO
IUPAC-namn (2R)-N-[3-[2-[2-[3-[[(2R)-2,4-dihydroxy-3,3-dimethylbutanoyl]amino]propanoylamino]ethyldisulfanyl]ethylamino]-3-oxopropyl]-2,4-dihydroxy-3,3-dimethylbutanamide
Molekylvikt (g/mol) 554.72
Formel vikt 554.2444