Läs mer
D-Pantethine, TRC
Produktkod. 30420283
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30420283 500 mg
30408987 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30420283 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP205823500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material D-Pantethine
CAS 16816-67-4
Molekylformel C22 H42 N4 O8 S2
Kvantitet 500 mg
Synonym Butanamide','N,N'-[dithiobis[2,1-ethanediylimino(3-oxo-3,1-propanediyl)]]bis[2,4-dihydroxy-3,3-dimethyl-','(2R,2'R)-, Butanamide','N,N'-[dithiobis[2,1-ethanediylimino(3-oxo-3,1-propanediyl)]]bis[2,4-dihydroxy-3,3-dimethyl-','[R-(R*,R*)]-, Butyramide','N,N'-[dithiobis(ethyleneiminocarbonylethylene)]bis[2,4-dihydroxy-3,3-dimethyl-','D-(+)- (8CI), (2R,2'R)-N,N'-[Dithiobis[2,1-ethanediylimino(3-oxo-3,1-propanediyl)]]bis[2,4-dihydroxy-3,3-dimethylbutanamide], Bis(N-pantothenylamidoethyl) disulfide, Bis(pantothenamidoethyl) disulfide, D-Bis-pantothenyl-β-aminoethyl disulfide, D-Pantethine, LBF disulfide, Lipodel, Pantethine, Pantetina, Panthecin, Pantomin, Pantosin
InChI formel InChI=1S/C22H42N4O8S2/c1-21(2,13-27)17(31)19(33)25-7-5-15(29)23-9-11-35-36-12-10-24-16(30)6-8-26-20(34)18(32)22(3,4)14-28/h17-18,27-28,31-32H,5-14H2,1-4H3,(H,23,29)(H,24,30)(H,25,33)(H,26,34)/t17-,18-/m0/s1
LEDER CC(C)(CO)[C@@H](O)C(=O)NCCC(=O)NCCSSCCNC(=O)CCNC(=O)[C@H](O)C(C)(C)CO
IUPAC-namn (2R)-N-[3-[2-[2-[3-[[(2R)-2,4-dihydroxy-3,3-dimethylbutanoyl]amino]propanoylamino]ethyldisulfanyl]ethylamino]-3-oxopropyl]-2,4-dihydroxy-3,3-dimethylbutanamide
Molekylvikt (g/mol) 554.72
Formel vikt 554.2444