Läs mer
D-Pemetrexed Dimethyl Diester, TRC
Produktkod. 30423466
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30423466 10 mg
30416985 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30423466 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP21953510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1391068-12-4
Molekylformel C22H25N5O6
Molekylvikt (g/mol) 455.46
Synonym N-[4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-L-glutamic Acid 1,5-Dimethyl Diester
IUPAC-namn dimethyl (2R)-2-[[4-[2-(2-amino-4-oxo-1,7-dihydropyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]amino]pentanedioate
LEDER COC(=O)CC[C@@H](NC(=O)c1ccc(CCc2c[nH]c3NC(=NC(=O)c23)N)cc1)C(=O)OC
Specifikationer
Kemiskt namn eller material D-Pemetrexed Dimethyl Diester
Kvantitet 10 mg
InChI formel InChI=1S/C22H25N5O6/c1-32-16(28)10-9-15(21(31)33-2)25-19(29)13-6-3-12(4-7-13)5-8-14-11-24-18-17(14)20(30)27-22(23)26-18/h3-4,6-7,11,15H,5,8-10H2,1-2H3,(H,25,29)(H4,23,24,26,27,30)/t15-/m1/s1
Formel vikt 455.1805