Learn More
[D-Phe12, Leu14] Bombesin, Alfa Aesar™
For cellular and molecular biology applications
Brand:  Alfa Aesar J66013.#0
Additional Details : CAS Number : 108437-88-3 Weight : 0.00005kg
 View more versions of this product
Product Code. 15497199
  • 2316.90 SEK / 5mg
Estimated Shipment date
from Supplier 02-07-2020
Add to basket
Specifications
Specifications
1611.84
5mg
Glp-Gln-Arg-Leu-Gly-Asn-Gln-Trp-Ala-Val-Gly-DPhe-Leu-Leu-NH2
-30° to -10°C
[D-Phe12, Leu14] Bombesin
HPLC
Solid
RUO
Bombesin Receptor Antagonist
Solid
Videos
SDS Documents
For Research Use Only