Läs mer
D-Ribonolactone 2,3-Cyclohexyl Ketal, TRC
Produktkod. 30414921
Change view
Kvantitet:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30414921 500 mg
30426998 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30414921 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR416005500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material D-Ribonolactone 2,3-Cyclohexyl Ketal
CAS 27304-20-7
Molekylformel C11H16O5
Kvantitet 500 mg
Synonym 2,3-O-Cyclohexylidene-D-ribono-1,4-lactone, 2,3-O-Cyclohexylidene-D-ribonic Acid gamma-lactone
InChI formel InChI=1S/C11H16O5/c12-6-7-8-9(10(13)14-7)16-11(15-8)4-2-1-3-5-11/h7-9,12H,1-6H2/t7-,8-,9-/m1/s1
LEDER OC[C@H]1OC(=O)[C@@H]2OC3(CCCCC3)O[C@H]12
IUPAC-namn (3aR,6R,6aR)-6-(hydroxymethyl)spiro[6,6a-dihydro-3aH-furo[3,4-d][1,3]dioxole-2,1'-cyclohexane]-4-one
Molekylvikt (g/mol) 228.24
Formel vikt 228.0998