Läs mer
D-Ribopyranosylamine, TRC
Produktkod. 30417353
Change view
Kvantitet:
1 g
2.5 g
5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30417353 5 g -
30413340 1 g -
30413295 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30417353 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR4168505G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 43179-09-5
Molekylformel C5 H11 N O4
Molekylvikt (g/mol) 149.15
IUPAC-namn (2R,3R,4S,5R)-2-aminooxane-3,4,5-triol
LEDER N[C@@H]1OC[C@@H](O)[C@H](O)[C@H]1O
Specifikationer
Kemiskt namn eller material D-Ribopyranosylamine
Kvantitet 5 g
InChI formel InChI=1S/C5H11NO4/c6-5-4(9)3(8)2(7)1-10-5/h2-5,7-9H,1,6H2/t2-,3+,4-,5-/m1/s1
Formel vikt 149.0688