Läs mer
D-Ribose 1-Phosphate, Biscyclohexylammonium Salt, TRC
Produktkod. 30179316
Change view
Kvantitet:
1 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179316 10 mg
30179422 25 mg
30179507 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179316 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR49500010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 58459-37-3
Molekylformel C6 H13 N . C5 H11 O8 P
Molekylvikt (g/mol) 329.28
Synonym D-Ribofuranose, 1-(dihydrogen phosphate), compd. with cyclohexanamine (1:2), D-Ribose 1-phosphate biscyclohexylammonium salt, D-Ribose 1-phosphate bis(cyclohexylammonium) salt, D-Ribfuranose 1-phosphate bis(cyclohexylammonium) salt, D-Ribose 1-phosphate cyclohexylamine salt
IUPAC-namn cyclohexanamine;[(3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl] dihydrogen phosphate
LEDER NC1CCCCC1.OC[C@H]2OC(OP(=O)(O)O)[C@H](O)[C@@H]2O
Specifikationer
Kemiskt namn eller material D-Ribose 1-Phosphate, Biscyclohexylammonium Salt
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C6H13N.C5H11O8P/c7-6-4-2-1-3-5-6;6-1-2-3(7)4(8)5(12-2)13-14(9,10)11/h6H,1-5,7H2;2-8H,1H2,(H2,9,10,11)/t;2-,3-,4-,5?/m.1/s1
Formel vikt 329.124
Procent renhet >95%