Läs mer
D-Ribose-13C5, TRC
Produktkod. 30179627
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179627 100 mg
30179466 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30179627 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR417521100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 202114-47-4
Molekylformel 13C5H10O5
Molekylvikt (g/mol) 155.09
Synonym Ribose-13C5, D-(-)-Ribose-13C5, D-[UL-13C5]-Ribose;
IUPAC-namn (2R,3R,4R)-2,3,4,5-tetrahydroxy(1,2,3,4,5-^{13}C_{5})pentanal
LEDER O[13C@H]1[13C@@H](O)[13CH](O)O[13C@@H]1[13CH2]O
Specifikationer
Kemiskt namn eller material D-Ribose-13C5
Kvantitet 100 mg
InChI formel InChI=1S/C5H10O5/c6-1-3(8)5(10)4(9)2-7/h1,3-5,7-10H,2H2/t3-,4+,5-/m0/s1/i1+1,2+1,3+1,4+1,5+1
Formel vikt 155.0696