Läs mer
D-Sorbitol Polyglycidyl Ether (Technical Grade), TRC
Produktkod. 30422219
Change view
Kvantitet:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30422219 5 g
30428622 10 g
30417685 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30422219 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6771085G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 68412-01-1
Synonym D-Glucitol, reaction products with epichlorohydrin, D-Sorbitol Polyglycidyl Ether
LEDER OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)CO.ClCC1CO1
Specifikationer
Kemiskt namn eller material D-Sorbitol Polyglycidyl Ether (Technical Grade)
Kvantitet 5 g
Formel vikt 274.0819