Läs mer
D-Talose-1-13C, TRC
Produktkod. 30426608
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30426608 10 mg
30414248 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30426608 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT00556210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 70849-29-5
Molekylformel 13CC5H12O6
Molekylvikt (g/mol) 181.16
Synonym D(+)-Talose-13C, NSC 224293-13C
IUPAC-namn (2S,3S,4S,5R)-2,3,4,5,6-pentahydroxy(1^{13}C)hexanal
LEDER OC[C@@H](O)[C@H](O)[C@H](O)[C@H](O)[13CH]=O
Specifikationer
Kemiskt namn eller material D-Talose-1-13C
Kvantitet 10 mg
InChI formel InChI=1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h1,3-6,8-12H,2H2/t3-,4-,5-,6+/m1/s1/i1+1
Formel vikt 181.0667