missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAAM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25206-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
DAAM2 is a recently identified planar cell polarity (PCP) signaling molecule implicated in the regulation of cellular polarity, convergent extension, and invasion. DAAM2 belongs to the formin homology family and contains a DAD (diaphanous autoregulatory) domain, an FH1 (formin homology 1) domain, an FH2 (formin homology 2) domain and a GBD/FH3 (Rho GTPase-binding/ formin homology 3) domain. The DAAM2 protein is known to be involved in cell divison. Though the exact function of DAAM2 is yet unknown, reports suggest that it is closely related to DAAM1 and hence along with DAAM1 it may also be required in Wnt/Fz signaling and activation of Rho and in regulating cytoskeleton architecture. DAAM2 is expressed in most of tissues but the expression level ishigher in brain.Specifications
DAAM2 | |
Polyclonal | |
Unconjugated | |
DAAM2 | |
DAAM2, KIAA0381 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23500, 316201, 76441 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q80U19, Q86T65 | |
Daam2 | |
DAAM2 Fusion Protein Ag18955 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |