Läs mer
Dabigatran Ethyl Ester Hydrochloride Salt, TRC
Produktkod. 30136101
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136101 250 mg
30133495 100 mg
17992055 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136101 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD100135250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 211914-50-0
Molekylformel C27 H29 N7 O3 . Cl H
Molekylvikt (g/mol) 536.03
Synonym β-Alanine, N-[[2-[[[4-(aminoiminomethyl)phenyl]amino]methyl]-1-methyl-1H-benzimidazol-5-yl]carbonyl]-N-2-pyridinyl-, ethyl ester, monohydrochloride (9CI), Dabigatran Ethyl Ester Monohydrochlorid
IUPAC-namn ethyl 3-[[2-[(4-carbamimidoylanilino)methyl]-1-methylbenzimidazole-5-carbonyl]-pyridin-2-ylamino]propanoate;hydrochloride
LEDER Cl.CCOC(=O)CCN(C(=O)c1ccc2c(c1)nc(CNc3ccc(cc3)C(=N)N)n2C)c4ccccn4
Specifikationer
Kemiskt namn eller material Dabigatran ethyl ester hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C27H29N7O3.ClH/c1-3-37-25(35)13-15-34(23-6-4-5-14-30-23)27(36)19-9-12-22-21(16-19)32-24(33(22)2)17-31-20-10-7-18(8-11-20)26(28)29;/h4-12,14,16,31H,3,13,15,17H2,1-2H3,(H3,28,29);1H
Formel vikt 535.2099
Procent renhet >95%