Läs mer
Dabigatran, MedChemExpress
Produktkod. 16483468
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16483468 10 mg
16459956 50 mg
16426197 5 mg
16484656 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16483468 Leverantör Medchem Express Leverantörsnummer HY10163
MedChemExpress Dabigatran (BIBR 953), an oral anticoagulant, is a reversible, potent, competitive direct thrombin inhibitor (Ki=4.5 nM). Dabigatran (BIBR 953) also inhibits thrombin-induced platelet aggregation (IC50=10 nM).
Dabigatran (BIBR 953), an oral anticoagulant, is a reversible, potent, competitive direct thrombin inhibitor (Ki=4.5 nM). Dabigatran (BIBR 953) also inhibits thrombin-induced platelet aggregation (IC50=10 nM).
Specifikationer
Kemiskt namn eller material Dabigatran
CAS 211914-51-1
Färg Light Yellow
Anteckningar om renhetsgrad Research
Molekylformel C25H25N7O3
Kvantitet 10 mg
Synonym BIBR 953 BIBR 953ZW
Löslighetsinformation 0.1 M HCL : 12.5 mg/mL (26.51 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
LEDER N=C(N)C(C=C1)=CC=C1NCC2=NC3=CC(C(N(CCC(O)=O)C4=NC=CC=C4)=O)=CC=C3N2C
Molekylvikt (g/mol) 471.51
Formel vikt 471.51
Procent renhet 96.84%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation