missing translation for 'onlineSavingsMsg'
Learn More

Dabigatran, TRC

Product Code. 30008844
Change view
Click to view available options
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30008844 10 mg
30003594 1 mg
30009480 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008844 Supplier Toronto Research Chemicals Supplier No. TRCD10009010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 211914-51-1
Molekylformel C25 H25 N7 O3
Molekylvikt (g/mol) 471.51
Synonym N-[[2-[[[4-(aminoiminomethyl)phenyl]amino]methyl]-1-methyl-1H-benzimidazol-5-yl]carbonyl]-N-2-pyridinyl-β-Alanine, BIBR 953, BIBR 953ZW, Dabigatran, N-[[2-[[[4-(Aminoiminomethyl)phenyl]amino]methyl]-1-methyl-1H-benzimidazol-5-yl]carbonyl]-N-2-pyridinyl-β-alanine
IUPAC-namn 3-[[2-[(4-carbamimidoylanilino)methyl]-1-methylbenzimidazole-5-carbonyl]-pyridin-2-ylamino]propanoic acid
LEDER Cn1c(CNc2ccc(cc2)C(=N)N)nc3cc(ccc13)C(=O)N(CCC(=O)O)c4ccccn4

Specifications

Kemiskt namn eller material Dabigatran
Kvantitet 10 mg
InChI formel InChI=1S/C25H25N7O3/c1-31-20-10-7-17(25(35)32(13-11-23(33)34)21-4-2-3-12-28-21)14-19(20)30-22(31)15-29-18-8-5-16(6-9-18)24(26)27/h2-10,12,14,29H,11,13,15H2,1H3,(H3,26,27)(H,33,34)
Formel vikt 471.2019
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.