Läs mer
Dabigatran, TRC
Produktkod. 30009480
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009480 5 mg
30008844 10 mg
30003594 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009480 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD1000905MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 211914-51-1
Molekylformel C25 H25 N7 O3
Molekylvikt (g/mol) 471.51
Synonym N-[[2-[[[4-(aminoiminomethyl)phenyl]amino]methyl]-1-methyl-1H-benzimidazol-5-yl]carbonyl]-N-2-pyridinyl-β-Alanine, BIBR 953, BIBR 953ZW, Dabigatran, N-[[2-[[[4-(Aminoiminomethyl)phenyl]amino]methyl]-1-methyl-1H-benzimidazol-5-yl]carbonyl]-N-2-pyridinyl-β-alanine
IUPAC-namn 3-[[2-[(4-carbamimidoylanilino)methyl]-1-methylbenzimidazole-5-carbonyl]-pyridin-2-ylamino]propanoic acid
LEDER Cn1c(CNc2ccc(cc2)C(=N)N)nc3cc(ccc13)C(=O)N(CCC(=O)O)c4ccccn4
Specifikationer
Kemiskt namn eller material Dabigatran
Kvantitet 5 mg
InChI formel InChI=1S/C25H25N7O3/c1-31-20-10-7-17(25(35)32(13-11-23(33)34)21-4-2-3-12-28-21)14-19(20)30-22(31)15-29-18-8-5-16(6-9-18)24(26)27/h2-10,12,14,29H,11,13,15H2,1H3,(H3,26,27)(H,33,34)
Formel vikt 471.2019