Läs mer
DACA, TRC
Produktkod. 30355579
Change view
Kvantitet:
2.5 mg
5 mg
10 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30355579 10 mg -
30355507 2.5 mg 2.5mg
30354720 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30355579 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD10120010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 89459-25-6
Molekylformel C18H19N3O
Molekylvikt (g/mol) 293.36
Synonym N-[2-(Dimethylamino)ethyl]-4-acridinecarboxamide, DACA, N-[2-(Dimethylamino)ethyl]acridine-4-carboxamide, NSC 601316
IUPAC-namn N-[2-(dimethylamino)ethyl]acridine-4-carboxamide
LEDER OC(=O)CC1CCC2(CC1)OOC3(OO2)C4CC5CC3CC(C5)C4
Specifikationer
Kemiskt namn eller material DACA
Kvantitet 10 mg
InChI formel InChI=1S/C18H19N3O/c1-21(2)11-10-19-18(22)15-8-5-7-14-12-13-6-3-4-9-16(13)20-17(14)15/h3-9,12H,10-11H2,1-2H3,(H,19,22)