missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ DALRD3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557850
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Immunogen sequence: LQEALWHWPE DSHPGLAGAS DTGTGGCLVV HVVSCEEEFQ QQKLDLLWRK LVDKAPLRQK HLICGPVKVA GAPGTLMTAP EYYEFR Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 69%.
The exact function of this gene is not known. It encodes a protein with a DALR anticodon binding domain similar to that of class Ia aminoacyl tRNA synthetases. This gene is located in a cluster of genes (with a complex sense-anti-sense genome architecture) on chromosome 3, and contains two micro RNA (miRNA) precursors (mir-425 and mir-191) in one of its introns. Preferential expression of this gene (the miRNAs and other genes in the cluster) in testis suggests a role of this gene in spermatogenesis.
Specifikationer
| DALRD3 | |
| Polyclonal | |
| Unconjugated | |
| DALRD3 | |
| DALR anticodon binding domain containing 3; DALR anticodon-binding domain-containing protein 3; DALRD3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55152 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5D0E6 | |
| DALRD3 | |
| Recombinant protein corresponding to Human DALRD3. Recombinant protein control fragment (Product #RP-96496). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering