Läs mer
DAMPA, TRC
Produktkod. 30138036
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138036 500 mg
17984935 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138036 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD136500500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 19741-14-1
Molekylformel C15 H15 N7 O2
Molekylvikt (g/mol) 325.33
Synonym Benzoic acid, 4-[[(2,4-diamino-6-pteridinyl)methyl]methylamino]-, Benzoic acid, p-[[(2,4-diamino-6-pteridinyl)methyl]methylamino]- (6CI,8CI), 4-[[(2,4-Diamino-6-pteridinyl)methyl]methylamino]benzoic acid, 2,4-Diamino-N10-methylpteroic acid, 4-Amino-4-deoxy-10-methylpteroic acid, 4-Amino-4-deoxy-N10-methylpteroic acid, 4-Amino-4-deoxy-N10-methylpteroic acid, 4-Deoxy-4-amino-N10-methylpteroic acid, 4-[N-[(2,4-Diaminopyrimido[4,5-b]pyrazin-6-yl)methyl]-N-methylamino]benzoic acid, NSC 131463, NSC 133723, p-[[(2,4-Diamino-6-pteridinyl)methyl]methylamino]benzoic acid, 4-[[(2,4-Diaminopteridin-6-yl)methyl]methylamino]benzoic Acid, 4-Amino-N10-methylpteroic Acid, APA
IUPAC-namn 4-[(2,4-diaminopteridin-6-yl)methyl-methylamino]benzoic acid
LEDER CN(Cc1cnc2nc(N)nc(N)c2n1)c3ccc(cc3)C(=O)O
Specifikationer
Kemiskt namn eller material DAMPA
Anteckningar om renhetsgrad HPLC
Kvantitet 500 mg
InChI formel InChI=1S/C15H15N7O2/c1-22(10-4-2-8(3-5-10)14(23)24)7-9-6-18-13-11(19-9)12(16)20-15(17)21-13/h2-6H,7H2,1H3,(H,23,24)(H4,16,17,18,20,21)
Formel vikt 325.1287
Procent renhet >95%