missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ DANI Liners

Provides better analysis for a wide range of compounds. Trajan Scientific and Medical DANI Liners has unique deactivation process for SGE standard inlets.

Brand:  Trajan Scientific and Medical™ 092503

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15239043

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance


Description

Description

  • Each batch of liners tested for activity
  • Manufactured from borosilicate glass
  • Color coded by geometry for easy identification
  • For use with DANI instruments
Specifications

Specifications

Straight-through Liner with quartz wool
5/Pk.
Deactivated Liners For DANI GCs
6 mm
DANI Liners
Straight Liner with Wool for DANI
2 mm
70 mm
Videos