missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ DANI Liners

Provides better analysis for a wide range of compounds. Trajan Scientific and Medical DANI Liners has unique deactivation process for SGE standard inlets.

Brand:  Trajan Scientific and Medical™ 092505

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15259043

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance


Description

Description

  • Each batch of liners tested for activity
  • Manufactured from borosilicate glass
  • Color coded by geometry for easy identification
  • For use with DANI instruments
Specifications

Specifications

Single taper with quartz wool
5/Pk.
Deactivated Liners For DANI GCs
6mm
DANI Liners
Single taper liner with wool for DANI
4mm
70mm
Videos