Beskrivning
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
- Free expert technical support
Specifikationer
Specifikationer
| Förpackning | Glass Bottle |
| CAS | 1329642-60-5 |
| Fysisk form | Solid |
| Industry Type | Antibiotics |
| Kvantitet | 10 mg |
| Kvalitet | Analytical Standard |