Läs mer
Dansyl-dimethylamine, TRC
Produktkod. 30363877
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30363877 25 mg
30358524 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30363877 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD17850025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6059-62-7
Molekylformel C14H18N2O2S
Molekylvikt (g/mol) 278.37
Synonym 5-(Dimethylamino)-N,N-dimethyl-1-Naphthalenesulfonamide, DNS-dimethylamine, N,N-Dimethyl-5-(dimethylamino)naphthalene-1-sulfonamide
IUPAC-namn 5-(dimethylamino)-N,N-dimethylnaphthalene-1-sulfonamide
LEDER CN(C)c1cccc2c(cccc12)S(=O)(=O)N(C)C
Specifikationer
Kemiskt namn eller material Dansyl-dimethylamine
Kvantitet 25 mg
InChI formel InChI=1S/C14H18N2O2S/c1-15(2)13-9-5-8-12-11(13)7-6-10-14(12)19(17,18)16(3)4/h5-10H,1-4H3
Formel vikt 278.1089