missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAOA Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24692-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein that is an activator of the FAD-dependent enzyme D-amino acid oxidase, which degrades the gliotransmitter D-serine, a potent activator of N-methyl-D-aspartate (NMDA) type glutamate receptors. Polymorphisms in this gene have been implicated in susceptibility to schizophrenia and bipolar affective disorder, possibly due to decreased levels of D-serine and decreased NMDA receptor functioning. Alternatively spliced transcript variants encoding different isoforms have been identified.Specifications
DAOA | |
Polyclonal | |
Unconjugated | |
DAOA | |
D amino acid oxidase activator, DAOA, G72, LG72, Protein G72, SG72 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
267012 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P59103 | |
DAOA | |
DAOA Fusion Protein Ag19143 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |