Läs mer
Dapagliflozin Methyl Acetate, TRC
Produktkod. 30366915
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30366915 50 mg
1 options
Produktkod. 30366915 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD18361050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dapagliflozin Methyl Acetate
CAS 1632287-34-3
Molekylformel C23 H27 Cl O7
Kvantitet 50 mg
Synonym ((2R,3S,4R,5R,6S)-6-(4-Chloro-3-(4-ethoxybenzyl)phenyl)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)methyl Acetate, Dapagliflozin Impurity-S2D(Mono acetylated)
InChI formel InChI=1S/C23H27ClO7/c1-3-29-17-7-4-14(5-8-17)10-16-11-15(6-9-18(16)24)23-22(28)21(27)20(26)19(31-23)12-30-13(2)25/h4-9,11,19-23,26-28H,3,10,12H2,1-2H3/t19-,20-,21+,22-,23+/m1/s1
LEDER CCOc1ccc(Cc2cc(ccc2Cl)[C@@H]3O[C@H](COC(=O)C)[C@@H](O)[C@H](O)[C@H]3O)cc1
IUPAC-namn [(2R,3S,4R,5R,6S)-6-[4-chloro-3-[(4-ethoxyphenyl)methyl]phenyl]-3,4,5-trihydroxyoxan-2-yl]methyl acetate
Molekylvikt (g/mol) 450.909
Formel vikt 450.145