missing translation for 'onlineSavingsMsg'
Learn More

DAPK1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 25136-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16823685

  • 1236.20 SEK / 20µL
Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Death-associated protein kinase 1 is a positive mediator of gamma-interferon induced programmed cell death. DAPK1 encodes a structurally unique 160-kD calmodulin dependent serine-threonine kinase that carries 8 ankyrin repeats and 2 putative P-loop consensus sites. It is a tumor suppressor candidate.
Specifications

Specifications

DAPK1
1.01 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P53355
DAP kinase 1, DAPK, DAPK1
Rabbit
IgG
20 μL
-20°C
Primary
1612
Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
DAPK1
Liquid
DAPK1
DAPK1 Fusion Protein Ag18387
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers