missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DAPK3 (ZIPK), GST Tag Recombinant Protein
Human DAPK3 (ZIPK), GST Tag Recombinant Protein
Brand: Invitrogen PV3686
Additional Details : Weight : 0.05250kg
Description
DAPK3 is a serine⁄threonine kinase which is functionally linked to cell motility, smooth muscle contractility and cell death processes.Specifications
NP_001339 | |
1613, 7850 | |
10 μg | |
DAPK Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: CAMK Group | |
Z'-LYTE™, Omnia™ Bulk Substrate | |
death-associated protein kinase 3 | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
Kinase Assay | |
80 kDa | |
DAPK3 (ZIPK) | |
Human | |
GST Tag | |
Biochemical phospho-specific activity (no antibody) | |
Dry Ice | |
DAPK3 | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |