Läs mer
Dapoxetine N-Oxide (90%), TRC
Produktkod. 30360284
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30360284 2.5 mg 2.5mg
30365373 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30360284 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD1857102.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1346603-24-4
Molekylformel C21 H23 N O2
Molekylvikt (g/mol) 321.41
Synonym Dapoxetine N-Oxide
IUPAC-namn (1S)-N,N-dimethyl-3-naphthalen-1-yloxy-1-phenylpropan-1-amine oxide
LEDER C[N+](C)([O-])[C@@H](CCOc1cccc2ccccc12)c3ccccc3
Specifikationer
Kemiskt namn eller material Dapoxetine N-Oxide
Kvantitet 2.5 mg
InChI formel InChI=1S/C21H23NO2/c1-22(2,23)20(18-10-4-3-5-11-18)15-16-24-21-14-8-12-17-9-6-7-13-19(17)21/h3-14,20H,15-16H2,1-2H3/t20-/m0/s1
Formel vikt 321.1729