Läs mer
Dapoxetine, TRC
Produktkod. 30354118
Change view
Kvantitet:
1 mg
2 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30354118 1 mg
30354888 10 mg
30358186 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30354118 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD1857131MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 119356-77-3
Molekylformel C21H23NO
Molekylvikt (g/mol) 305.41
Synonym (alphaS)-N,N-Dimethyl-alpha-[2-(1-naphthalenyloxy)ethyl]benzenemethanamine, (S)-(+)-N,N-Dimethyl-3-(naphthalen-1-yloxy)-1-phenylpropan-1-amine, (S)-(+)-N,N-Dimethyl-alpha-[2-(1-naphthalenyloxy)ethyl]benzenemethanamine
IUPAC-namn (1S)-N,N-dimethyl-3-naphthalen-1-yloxy-1-phenylpropan-1-amine
LEDER CN(C)[C@@H](CCOc1cccc2ccccc12)c3ccccc3
Specifikationer
Kemiskt namn eller material Dapoxetine
Kvantitet 1 mg
InChI formel InChI=1S/C21H23NO/c1-22(2)20(18-10-4-3-5-11-18)15-16-23-21-14-8-12-17-9-6-7-13-19(17)21/h3-14,20H,15-16H2,1-2H3/t20-/m0/s1
Formel vikt 305.178