missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAPP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14722-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
May act as a B-cell-associated adapter that regulates B-cell antigen receptor (BCR)-signaling downstream of PI3K.Specifications
DAPP1 | |
Polyclonal | |
Unconjugated | |
DAPP1 | |
BAM32, DAPP1, hDAPP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26377, 27071, 362046 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9QXT1, Q9UN19 | |
Dapp1 | |
DAPP1 Fusion Protein Ag6459 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |