Läs mer
Dapsone, TRC
Produktkod. 17981535
Change view
Kvantitet:
1 g
10 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17981535 10 g
30137498 100 g
17950945 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17981535 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD19325010G
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 80-08-0
Kemiskt namn eller material Dapsone
Formel vikt 248.0619
InChI formel InChI=1S/C12H12N2O2S/c13-9-1-5-11(6-2-9)17(15,16)12-7-3-10(14)4-8-12/h1-8H,13-14H2
IUPAC-namn 4-(4-aminophenyl)sulfonylaniline
Molekylformel C12 H12 N2 O2 S
Molekylvikt (g/mol) 248.3
Procent renhet >95%
Anteckningar om renhetsgrad HPLC
LEDER Nc1ccc(cc1)S(=O)(=O)c2ccc(N)cc2
Synonym Aniline, 4,4'-sulfonyldi- (7CI,8CI), 4,4'-Sulfonylbis[benzenamine], 1,1'-Sulfonylbis[4-aminobenzene], 1358F, 4,4'-DDS, 4,4'-Dapsone, 4,4'-Diaminobiphenyl sulfone, 4,4'-Diaminodiphenyl sulfone, 4,4'-Sulfonylbisaniline, 4,4'-Sulfonylbisbenzamine, 4,4'-Sulfonyldianiline, 4,4'-Sulfonyldiphenylamine, 4,4'-Sulphonyldianiline, 4-(4-Aminophenyl)sulfonylaniline, 4-(4-Aminophenylsulfonyl)benzenamine, 4-Aminophenyl sulfone, Aczone, Aradur 9664-1, Aradur 9664-1NL, Aradur 976, Aradur 976-1, Aradur 9761-1, Araldite HT 976, Atrisone, Avlosulfon, Avlosulfone, Bis(4-aminophenyl) sulfone, Bis(p-aminophenyl) sulfone, Croysulfone, D 0089, DADFS, DADPS, DDS, DDS (pharmaceutical), DSS, Dapson, Dapsone, Di(4-aminophenyl)sulfone, Di(p-aminophenyl) sulfone, Diaphenylsulfon, Diaphenylsulfone, Diphenasone, Diphone, Disulone, Dumitone, Eporal, F 1358, HB-MC-GE 40, HB-MC-GE 45, HEW 62, HT 9664, HT 976, HT 976-1, HY 976, Hardener HT 976, Lapox K 10, Metabolite C, NSC 6091, Novophone, Seikacure S, Servidapson, Sulfona, Sulfona-Mae, Sulphadione, Sumicure S, Udolac, p,p'-Diaminodiphenyl sulfone, p,p'-Sulfonyldianiline, p-Aminophenyl sulfone
Visa mer Visa mindre
Specifikationer
Kvantitet 10 g