Läs mer
Darbufelone mesylate, MedChemExpress
Produktkod. 16486087
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16486087 5 mg
16443368 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16486087 Leverantör Medchem Express Leverantörsnummer HY101438A
MedChemExpress Darbufelone mesylate (CI-1004 mesylate) is a dual inhibitor of cellular PGF2α and LTB4 production. Darbufelone potently inhibits PGHS-2 (IC50 = 0.19 μM) but is much less potent with PGHS-1 (IC50= 20 μM).
Darbufelone mesylate (CI-1004 mesylate) is a dual inhibitor of cellular PGF2α and LTB4 production. Darbufelone potently inhibits PGHS-2 (IC50 = 0.19 μM) but is much less potent with PGHS-1 (IC50= 20 μM).
Specifikationer
Kemiskt namn eller material Darbufelone mesylate
CAS 139340-56-0
Färg Light Yellow
Anteckningar om renhetsgrad Research
Molekylformel C19H28N2O5S2
Kvantitet 5 mg
Synonym CI-1004 mesylate
Löslighetsinformation DMSO : 110 mg/mL (256.67 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
LEDER O=C1N=C(N)S/C1=C\C2=CC(C(C)(C)C)=C(O)C(C(C)(C)C)=C2.CS(=O)(O)=O
Molekylvikt (g/mol) 428.57
Formel vikt 428.57
Procent renhet 98.45%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Metabolism-protein/nucleotide metabolism