Läs mer
Darinaparsin, TRC
Produktkod. 30362616
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30362616 500 mg
30359720 50 mg
30353937 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30362616 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD193425500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 69819-86-9
Molekylformel C12H22AsN3O6S
Molekylvikt (g/mol) 411.31
Synonym N-[S-(Dimethylarsino)-N-L-γ-glutamyl-L-cysteinyl]-glycine, Dimethylarsinic Glutathione, ZIO 101, Zinapar, L-γ-Glutamyl-S-(dimethylarsino)-L-cysteinyl-glycine
IUPAC-namn (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-3-dimethylarsanylsulfanyl-1-oxopropan-2-yl]amino]-5-oxopentanoic acid
LEDER CC/C=C\C/C=C\C/C=C\CCCCCCCC(=O)OCC1OC1
Specifikationer
Kemiskt namn eller material Darinaparsin
Kvantitet 500 mg
InChI formel InChI=1S/C12H22AsN3O6S/c1-13(2)23-6-8(11(20)15-5-10(18)19)16-9(17)4-3-7(14)12(21)22/h7-8H,3-6,14H2,1-2H3,(H,15,20)(H,16,17)(H,18,19)(H,21,22)/t7-,8-/m0/s1
Formel vikt 411.0445