Läs mer
Dasatinib Carboxylic Acid Ethyl Ester, TRC
Produktkod. 30352457
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30352457 25 mg
1 options
Produktkod. 30352457 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD19361525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dasatinib Carboxylic Acid Ethyl Ester
CAS 910297-62-0
Molekylformel C24H28ClN7O3S
Kvantitet 25 mg
Synonym 4-[6-[[5-[[(2-Chloro-6-methylphenyl)amino]carbonyl]-2-thiazolyl]amino]-2-methyl-4-pyrimidinyl]-1-piperazineacetic Acid Ethyl Ester
InChI formel InChI=1S/C24H28ClN7O3S/c1-4-35-21(33)14-31-8-10-32(11-9-31)20-12-19(27-16(3)28-20)29-24-26-13-18(36-24)23(34)30-22-15(2)6-5-7-17(22)25/h5-7,12-13H,4,8-11,14H2,1-3H3,(H,30,34)(H,26,27,28,29)
LEDER O=C(C1=CN=C(NC2=CC(N3CCN(CC(OCC)=O)CC3)=NC(C)=N2)S1)NC4=C(C)C=CC=C4Cl
IUPAC-namn ethyl 2-(4-(6-((5-((2-chloro-6-methylphenyl)carbamoyl)thiazol-2-yl)amino)-2-methylpyrimidin-4-yl)piperazin-1-yl)acetate
Molekylvikt (g/mol) 530.04
Formel vikt 529.17