Läs mer
Daunomycinone, TRC
Produktkod. 30356794
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30356794 50 mg
1 options
Produktkod. 30356794 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD19385050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Daunorubicinone
CAS 21794-55-8
Molekylformel C21 H18 O8
Kvantitet 50 mg
Synonym (8S,10S)-8-Acetyl-7,8,9,10-tetrahydro-6,8,10,11-tetrahydroxy-1-methoxy-5,12-naphthacenedione, (8S-cis)-8-Acetyl-7,8,9,10-tetrahydro-6,8,10,11-tetrahydroxy-1-methoxy-5,12-naphthacenedione, Daunomycinone, (+)-Daunomycinone, Daunomycin aglicone, Daunomycinon, Daunorubicin aglycone, Daunorubicinone, Leukaemomycinone C, NSC 109351
InChI formel InChI=1S/C21H18O8/c1-8(22)21(28)6-10-13(11(23)7-21)19(26)16-15(18(10)25)17(24)9-4-3-5-12(29-2)14(9)20(16)27/h3-5,11,23,25-26,28H,6-7H2,1-2H3/t11-,21-/m0/s1
LEDER COc1cccc2C(=O)c3c(O)c4C[C@](O)(C[C@H](O)c4c(O)c3C(=O)c12)C(=O)C
IUPAC-namn (7S,9S)-9-acetyl-6,7,9,11-tetrahydroxy-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione
Molekylvikt (g/mol) 398.36
Formel vikt 398.1002