Learn More
Emma Safety Footwear Dave (Ruffneck) Safety Shoes
Looks and feels like a classic sneaker. Highly supportive reinforced ruff neck heel together with the shock absorbing sole keeps ankles, shins and joints free from stress and injury.
Brand: Emma Safety Footwear 914546-42
Additional Details : Weight : 1.58000kg
Description
- S3-Safety shoe with sole plate for wet work
- Nubuck / Oil Nubuck construction
- GripForce™ Easy Twist Outsole
- 3M™ Reflective Striping
- ESD certified
- Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifications
EN ISO 20345:2011, EN 12568:2010 | |
Brown | |
Hydro-Tec™ Sanitized Silver | |
42 |
Laces | |
Dave (Ruffneck) | |
Nubuck, Oiled Nubuck | |
Steel |