Learn More
Masterflex™ DB15 Male Connector
Product Code. 10739484
Click to view available options
For Use With:
Masterflex™ Drives
Unit Size:
Each
Product Code. 10739484
Brand: Masterflex™ 0759552
Only null left
Add to basket
Add to basket
Masterflex™ DB15 Male Connector to create your own cable for optimization of Masterflex(R) drives
  • Combine connectors with 20-22 gauge shielded wire to create your own interface cable
  • Maximum cable length is 300 m
Specifications
For Use With (Application) Creating Your Own Cable
For Use With Masterflex™ Drives
Inlet Connection (English) DB15
Type Male Connector