missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DBC1 Mouse anti-Human, Mouse, Porcine, Clone: 2A10E3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60336-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The function of this protein remains unknown.Specifications
DBC1 | |
Monoclonal | |
1.57 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60477, Q920P3 | |
Brinp1 | |
DBC1 Fusion Protein Ag6588 | |
20 μL | |
Primary | |
Mouse, Human, Porcine | |
Antibody | |
IgG2c |
Immunofluorescence, Western Blot, Immunocytochemistry | |
2A10E3 | |
Unconjugated | |
BRINP1 | |
BRINP1, DBC1, DBCCR1, deleted in bladder cancer 1, FAM5A, Protein FAM5A | |
Mouse | |
Protein A | |
RUO | |
100625235, 1620, 56710 | |
-20°C | |
Liquid |