missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DBC1 Mouse anti-Human, Mouse, Porcine, Clone: 2A10E3, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | DBC1 |
---|---|
Clone | 2A10E3 |
Concentration | 1.57 mg/mL |
Applications | Immunofluorescence, Western Blot, Immunocytochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16827445
|
Proteintech
60336-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 19-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16817445
|
Proteintech
60336-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 19-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The function of this protein remains unknown.Specifications
DBC1 | |
1.57 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
BRINP1, DBC1, DBCCR1, deleted in bladder cancer 1, FAM5A, Protein FAM5A | |
Brinp1 | |
IgG2c | |
Protein A | |
Antibody |
2A10E3 | |
Immunofluorescence, Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Mouse, Human, Porcine | |
O60477, Q920P3 | |
100625235, 1620, 56710 | |
DBC1 Fusion Protein Ag6588 | |
Primary | |
-20°C | |
BRINP1 |