Läs mer
DBCO-PEG5-NHS ester, TRC
Produktkod. 30355288
Change view
Kvantitet:
1 mg
2 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30355288 10 mg
30364764 1 mg
30351191 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30355288 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD19499810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1378531-80-6
Molekylformel C36H43N3O11
Specifikationer
Certifikat och efterlevnad ISO 9001
Molekylvikt (g/mol) 693.74